Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122035.3 | NP_197528.1 | 8 - 9 | 1944 | 96 | |
GenBank mRNA | AY039911.1 | - | 8 - 9 | 1770 | 96 | |
AY079334.1 | - | 8 - 9 | 1704 | 96 | ||
KJ138676.1 | - | 8 - 9 | 1704 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |