Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203419.2 | NP_001190348.1 | 20 - 21 | 4185 | 146 | |
NM_001343652.1 | NP_001330078.1 | 17 - 18 | 3353 | 146 | ||
NM_122039.5 | NP_197532.3 | 20 - 21 | 4186 | 146 | ||
GenBank mRNA | DQ118423.1 | - | 20 - 21 | 4186 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |