Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161252.2 | NP_001154724.1 | 14 - 15 | 1672 | 94 | |
NM_001343670.1 | NP_001318611.1 | 14 - 15 | 1651 | 94 | ||
NM_001343671.1 | NP_001318612.1 | 15 - 16 | 2104 | 94 | ||
GenBank mRNA | HQ427882.1 | - | 14 - 15 | 1654 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |