Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122059.4 | NP_568395.1 | 2 - 3 | 474 | 77 | |
GenBank mRNA | AB182157.1 | - | 2 - 3 | 394 | 77 | |
AY080751.1 | - | 2 - 3 | 409 | 77 | ||
AY087569.1 | - | 2 - 3 | 405 | 77 | ||
AY114009.1 | - | 2 - 3 | 275 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |