Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343688.1 | NP_001332342.1 | 5 - 6 | 642 | 83 | |
NM_122092.3 | NP_568402.1 | 5 - 6 | 640 | 83 | ||
GenBank mRNA | U43528.1 | - | 5 - 6 | 584 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |