Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085142.1 | NP_001078611.1 | 10 - 11 | 1875 | 118 | |
NM_122107.4 | NP_197598.2 | 10 - 11 | 1928 | 118 | ||
GenBank mRNA | AK229291.1 | - | 10 - 11 | 1875 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |