Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122138.3 | NP_197625.1 | 8 - 9 | 1229 | 148 | |
GenBank mRNA | AY084606.1 | - | 8 - 9 | 1143 | 148 | |
BT000923.1 | - | 8 - 9 | 1020 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |