Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122181.3 | NP_197667.1 | 14 - 15 | 2457 | 116 | |
GenBank mRNA | AK228695.1 | - | 14 - 15 | 2431 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |