Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036849.2 | NP_001031926.1 | 5 - 6 | 532 | 78 | |
NM_122214.4 | NP_197699.2 | 5 - 6 | 619 | 78 | ||
GenBank mRNA | AK118129.1 | - | 5 - 6 | 527 | 78 | |
AY518689.1 | - | 5 - 6 | 488 | 78 | ||
BT008588.1 | - | 5 - 6 | 488 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |