Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343791.1 | NP_001330849.1 | 6 - 7 | 3083 | 89 | |
NM_001343792.1 | NP_001330848.1 | 6 - 7 | 2991 | 89 | ||
NM_122221.3 | NP_197706.1 | 6 - 7 | 2991 | 89 | ||
GenBank mRNA | AB493755.1 | - | 6 - 7 | 2653 | 89 | |
AF116556.1 | - | 6 - 7 | 2991 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |