Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343792.1 | NP_001330848.1 | 9 - 10 | 4348 | 68 | |
NM_122221.3 | NP_197706.1 | 9 - 10 | 4348 | 68 | ||
GenBank mRNA | AB493755.1 | - | 9 - 10 | 4010 | 68 | |
AF116556.1 | - | 9 - 10 | 4348 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |