Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122236.3 | NP_568428.1 | 9 - 10 | 1471 | 83 | |
GenBank mRNA | AF454729.1 | - | 9 - 10 | 1310 | 83 | |
AK228165.1 | - | 9 - 10 | 1471 | 83 | ||
BT026524.1 | - | 9 - 10 | 1310 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |