Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343836.1 | NP_001332637.1 | 3 - 4 | 900 | 87 | |
NM_122308.5 | NP_197791.1 | 3 - 4 | 1030 | 87 | ||
GenBank mRNA | BT015364.1 | - | 3 - 4 | 862 | 87 | |
BT015721.1 | - | 2 - 3 | 720 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |