Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343841.1 | NP_001330409.1 | 3 - 4 | 1081 | 93 | |
NM_001343842.1 | NP_001330408.1 | 3 - 4 | 1154 | 93 | ||
NM_122317.4 | NP_197800.1 | 3 - 4 | 1081 | 93 | ||
GenBank mRNA | AK316888.1 | - | 3 - 4 | 966 | 93 | |
Y18550.1 | - | 860 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |