Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122491.3 | NP_197962.1 | 5 - 6 | 1103 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |