Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122564.5 | NP_198034.2 | 2 - 3 | 672 | 105 | |
GenBank mRNA | AY062606.1 | - | 2 - 3 | 624 | 105 | |
AY128787.1 | - | 2 - 3 | 383 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |