Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102123.4 | NP_173690.1 | 1 - 2 | 577 | 75 | |
GenBank mRNA | AY054490.1 | - | 1 - 2 | 576 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |