Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102123.4 | NP_173690.1 | 4 - 5 | 1529 | 66 | |
GenBank mRNA | AY054490.1 | - | 4 - 5 | 1528 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |