Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102123.4 | NP_173690.1 | 6 - 7 | 2041 | 99 | |
GenBank mRNA | AY054490.1 | - | 6 - 7 | 2040 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |