Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102235.2 | NP_173799.1 | 2 - 3 | 2200 | 76 | |
GenBank mRNA | AY072210.1 | - | 2 - 3 | 2201 | 76 | |
AY096366.1 | - | 1 - 2 | 1981 | 76 | ||
KJ138988.1 | - | 1 - 2 | 1981 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |