Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332636.1 | NP_001320353.1 | 4 - 5 | 956 | 66 | |
NM_102254.3 | NP_173818.1 | 4 - 5 | 1011 | 66 | ||
GenBank mRNA | KJ676988.1 | - | 4 - 5 | 761 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |