Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332636.1 | NP_001320353.1 | 7 - 8 | 1939 | 97 | |
NM_102254.3 | NP_173818.1 | 8 - 9 | 1567 | 97 | ||
GenBank mRNA | KJ676988.1 | - | 8 - 9 | 1317 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |