Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102258.6 | NP_173822.2 | 5 - 6 | 873 | 73 | |
GenBank mRNA | AY074564.1 | - | 5 - 6 | 800 | 73 | |
AY226825.1 | - | 5 - 6 | 805 | 73 | ||
BT025997.1 | - | 5 - 6 | 557 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |