Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198151.2 | NP_001185080.1 | 19 - 20 | 3800 | 88 | |
NM_001332641.1 | NP_001323398.1 | 19 - 20 | 3756 | 88 | ||
NM_102265.3 | NP_173829.3 | 19 - 20 | 3737 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |