Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102290.4 | NP_173854.1 | 2 - 3 | 270 | 67 | |
GenBank mRNA | AY045598.1 | - | 2 - 3 | 212 | 67 | |
AY094027.1 | - | 2 - 3 | 166 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |