Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332663.1 | NP_001322233.1 | 9 - 10 | 2098 | 55 | |
NM_001332664.1 | NP_001322232.1 | 10 - 11 | 2077 | 55 | ||
NM_102311.7 | NP_173873.5 | 10 - 11 | 2018 | 55 | ||
GenBank mRNA | AY128287.1 | - | 10 - 11 | 1963 | 55 | |
BT000826.1 | - | 10 - 11 | 1864 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |