Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198165.1 | NP_001185094.1 | 6 - 7 | 564 | 84 | |
NM_102345.4 | NP_173906.2 | 6 - 7 | 609 | 84 | ||
GenBank mRNA | AY062594.1 | - | 6 - 7 | 563 | 84 | |
AY114653.1 | - | 6 - 7 | 508 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |