Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084127.1 | NP_001077596.1 | 8 - 9 | 1871 | 73 | |
NM_102365.5 | NP_173925.3 | 9 - 10 | 2057 | 73 | ||
GenBank mRNA | AY170377.1 | - | 9 - 10 | 1688 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |