Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332688.1 | NP_001322403.1 | 3 - 4 | 2055 | 106 | |
NM_102380.4 | NP_173940.2 | 3 - 4 | 2622 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |