Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CB5-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102398.3 | NP_173958.1 | 1 - 2 | 212 | 100 | |
GenBank mRNA | AF332415.1 | - | 1 - 2 | 84 | 100 | |
AK117459.1 | - | 1 - 2 | 170 | 100 | ||
AY084761.1 | - | 1 - 2 | 176 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |