Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332720.1 | NP_001323415.1 | 1 | 757 | 106 | |
NM_102422.3 | NP_564251.1 | 1 | 654 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |