Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102494.4 | NP_564276.1 | 10 - 11 | 4067 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |