Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102494.4 | NP_564276.1 | 7 - 8 | 2533 | 93 | |
GenBank mRNA | AB046870.1 | - | 7 - 8 | 1642 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |