Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102508.3 | NP_174064.1 | 3 - 4 | 998 | 57 | |
GenBank mRNA | BT022053.1 | - | 3 - 4 | 839 | 57 | |
BT025555.1 | - | 3 - 4 | 722 | 57 | ||
KJ138982.1 | - | 3 - 4 | 722 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |