Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332791.1 | NP_001322317.1 | 4 - 5 | 918 | 89 | |
NM_102557.3 | NP_174113.2 | 4 - 5 | 1211 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |