Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102563.4 | NP_174119.1 | 8 - 9 | 1283 | 100 | |
GenBank mRNA | AB175035.1 | - | 8 - 9 | 977 | 100 | |
AF360166.1 | - | 8 - 9 | 1036 | 100 | ||
AY113914.1 | - | 8 - 9 | 977 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |