Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084147.1 | NP_001077616.1 | 2 - 3 | 672 | 87 | |
NM_102594.3 | NP_174150.1 | 1 - 2 | 944 | 87 | ||
GenBank mRNA | BT029488.1 | - | 1 - 2 | 772 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |