Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036025.5 | NP_001031102.2 | 4 - 5 | 362 | 65 | |
NM_001332815.1 | NP_001321010.1 | 1 - 2 | 234 | 65 | ||
NM_102617.4 | NP_564310.1 | 4 - 5 | 480 | 65 | ||
GenBank mRNA | AF355754.2 | - | 3 - 4 | 284 | 65 | |
AY150420.1 | - | 3 - 4 | 284 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |