Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123901.2 | NP_001117373.2 | 7 - 8 | 776 | 96 | |
NM_001332819.1 | NP_001321583.1 | 5 - 6 | 826 | 96 | ||
NM_102624.4 | NP_174178.2 | 6 - 7 | 743 | 96 | ||
GenBank mRNA | AY099624.1 | - | 6 - 7 | 654 | 96 | |
BT003404.1 | - | 3 - 4 | 186 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |