Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123901.2 | NP_001117373.2 | 9 - 10 | 960 | 86 | |
NM_001332819.1 | NP_001321583.1 | 7 - 8 | 1010 | 86 | ||
NM_102624.4 | NP_174178.2 | 8 - 9 | 927 | 86 | ||
GenBank mRNA | AY099624.1 | - | 8 - 9 | 838 | 86 | |
BT003404.1 | - | 5 - 6 | 370 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |