Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102668.4 | NP_174222.2 | 1 - 2 | 392 | 79 | |
GenBank mRNA | AB493485.1 | - | 1 - 2 | 140 | 79 | |
AF452173.1 | - | 1 - 2 | 140 | 79 | ||
BT026354.1 | - | 1 - 2 | 140 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |