Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106026.3 | NP_565070.2 | 8 - 9 | 1681 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |