Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106062.4 | NP_177543.1 | 2 - 3 | 922 | 103 | |
GenBank mRNA | AY034978.1 | - | 2 - 3 | 722 | 103 | |
AY062968.1 | - | 2 - 3 | 691 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |