Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106062.4 | NP_177543.1 | 3 - 4 | 1085 | 117 | |
GenBank mRNA | AY034978.1 | - | 3 - 4 | 885 | 117 | |
AY062968.1 | - | 3 - 4 | 854 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |