Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334634.1 | NP_001322432.1 | 2 - 3 | 548 | 108 | |
NM_106077.4 | NP_177557.2 | 3 - 4 | 900 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |