Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344004.1 | NP_001330510.1 | 14 - 15 | 1445 | 72 | |
NM_001344005.1 | NP_001330511.1 | 15 - 16 | 1292 | 72 | ||
NM_001344006.1 | NP_001330508.1 | 15 - 16 | 1241 | 72 | ||
NM_001344007.1 | NP_001330509.1 | 12 - 13 | 1222 | 72 | ||
NM_122618.3 | NP_568494.1 | 15 - 16 | 1242 | 72 | ||
GenBank mRNA | AY026255.1 | - | 15 - 16 | 1139 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |