Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122643.3 | NP_198113.2 | 15 - 16 | 2631 | 87 | |
GenBank mRNA | AB493764.1 | - | 14 - 15 | 2400 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |