Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122643.3 | NP_198113.2 | 2 - 3 | 330 | 62 | |
GenBank mRNA | AB493764.1 | - | 1 - 2 | 99 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |