Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCH;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344019.1 | NP_001330469.1 | 3 - 4 | 645 | 123 | |
NM_001344020.1 | NP_001330468.1 | 3 - 4 | 645 | 123 | ||
NM_122644.5 | NP_198114.2 | 3 - 4 | 661 | 123 | ||
GenBank mRNA | AB051072.1 | - | 3 - 4 | 186 | 123 | |
AK118707.1 | - | 3 - 4 | 643 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |