Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036883.2 | NP_001031960.1 | 5 - 6 | 1051 | 79 | |
NM_001344058.1 | NP_001318668.1 | 2 - 3 | 1558 | 79 | ||
NM_001344059.1 | NP_001332384.1 | 3 - 4 | 1506 | 79 | ||
NM_001344060.1 | NP_001332382.1 | 4 - 5 | 1402 | 79 | ||
NM_001344061.1 | NP_001332383.1 | 4 - 5 | 1155 | 79 | ||
NM_001344062.1 | NP_001332380.1 | 5 - 6 | 978 | 79 | ||
NM_001344063.1 | NP_001332381.1 | 4 - 5 | 1047 | 79 | ||
NM_001344064.1 | NP_001332379.1 | 4 - 5 | 940 | 79 | ||
GenBank mRNA | AK228774.1 | - | 5 - 6 | 732 | 79 | |
BT015758.1 | - | 5 - 6 | 714 | 79 | ||
BT020173.1 | - | 3 - 4 | 464 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |